Product Details

SNP ID
rs80146633
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.18:22924006 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATTATAATAGGTGTTTCACTCCA[A/G]TCCTGACTGAGAAAGTCCAGTTTAT
Phenotype
MIM: 604124
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
MIR4741 PubMed Links

Gene Details

Gene
MIR4741
Gene Name
microRNA 4741
There are no transcripts associated with this gene.

Gene
RBBP8
Gene Name
RB binding protein 8, endonuclease
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002894.2 Intron NP_002885.1
NM_203291.1 Intron NP_976036.1
NM_203292.1 Intron NP_976037.1
XM_005258325.2 Intron XP_005258382.1
XM_005258326.3 Intron XP_005258383.1
XM_006722519.2 Intron XP_006722582.1
XM_006722520.2 Intron XP_006722583.1
XM_006722521.2 Intron XP_006722584.1
XM_011526132.2 Intron XP_011524434.1
XM_017025915.1 Intron XP_016881404.1
XM_017025916.1 Intron XP_016881405.1

View Full Product Details