Product Details

SNP ID
rs79048890
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57351678 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCCGCTTCTCATGGCAGCGGCGG[G/T]GCTGATGGACCGGGTTCAGGTGAGT
Phenotype
MIM: 613840
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF304 PubMed Links

Gene Details

Gene
ZNF304
Gene Name
zinc finger protein 304
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290318.1 405 Missense Mutation GGG,GTG G5V NP_001277247.1
NM_001290319.1 405 UTR 5 NP_001277248.1
NM_020657.3 405 Missense Mutation GGG,GTG G5V NP_065708.2
XM_011527145.2 405 UTR 5 XP_011525447.1
XM_017027024.1 405 UTR 5 XP_016882513.1

View Full Product Details