Product Details

SNP ID
rs78065887
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.20:45649681 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGGGCCCTCTCTGAGCATCCCAA[C/T]GATAAAACTCAGTCAACCCGGAGCA
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
WFDC11 PubMed Links

Gene Details

Gene
WFDC11
Gene Name
WAP four-disulfide core domain 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_147197.2 Intron NP_671730.1

View Full Product Details