Product Details

SNP ID
rs76772959
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:3797660 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTCAGGTAGGGACCCTGCTCTTCC[C/G]CAGCCGCAGCCGCCTGACGCACCTA
Phenotype
MIM: 116949
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CDC25B PubMed Links

Gene Details

Gene
CDC25B
Gene Name
cell division cycle 25B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001287516.1 407 Missense Mutation CCC,CGC P16R NP_001274445.1
NM_001287517.1 407 Intron NP_001274446.1
NM_001287518.1 407 Missense Mutation CCC,CGC P16R NP_001274447.1
NM_001287519.1 407 Intron NP_001274448.1
NM_001287520.1 407 UTR 5 NP_001274449.1
NM_001287522.1 407 UTR 5 NP_001274451.1
NM_001287524.1 407 Intron NP_001274453.1
NM_004358.4 407 Intron NP_004349.1
NM_021872.3 407 Missense Mutation CCC,CGC P80R NP_068658.1
NM_021873.3 407 Missense Mutation CCC,CGC P80R NP_068659.1

View Full Product Details