Product Details

SNP ID
rs76080446
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:41798842 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCTGCCTGGGTGGTACATGGTTG[C/T]TGTATTTTGCTATCTAGTTTCTTTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PRDM15 PubMed Links

Gene Details

Gene
PRDM15
Gene Name
PR domain 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040424.2 6106 UTR 3 NP_001035514.1
NM_001282934.1 6106 UTR 3 NP_001269863.1
NM_022115.4 6106 Intron NP_071398.3
XM_006724039.2 6106 UTR 3 XP_006724102.1
XM_006724040.2 6106 UTR 3 XP_006724103.1
XM_011529674.1 6106 UTR 3 XP_011527976.1
XM_011529675.1 6106 UTR 3 XP_011527977.1
XM_011529676.2 6106 UTR 3 XP_011527978.1
XM_011529677.2 6106 UTR 3 XP_011527979.1
XM_011529678.2 6106 UTR 3 XP_011527980.1
XM_011529679.2 6106 UTR 3 XP_011527981.1
XM_011529681.2 6106 UTR 3 XP_011527983.1
XM_011529683.1 6106 UTR 3 XP_011527985.1
XM_017028425.1 6106 UTR 3 XP_016883914.1
XM_017028426.1 6106 UTR 3 XP_016883915.1

View Full Product Details