Product Details

SNP ID
rs78948972
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:43152907 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTGAGGCAGGATGTCTGGCTGGAT[C/T]TACCAGAAAGTATTCACAGCGGGTT
Phenotype
MIM: 614479 MIM: 109610
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MCAT PubMed Links

Gene Details

Gene
MCAT
Gene Name
malonyl-CoA-acyl carrier protein transacylase
There are no transcripts associated with this gene.

Gene
TSPO
Gene Name
translocator protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000714.5 Intron NP_000705.2
NM_001256530.1 Intron NP_001243459.1
NM_001256531.1 Intron NP_001243460.1

View Full Product Details