Product Details

SNP ID
rs77313877
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:50739500 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTACGTGTAGGGATGCACTGAGGGA[A/G]GTCTTCAGAACGGCTCTTCTCAGAG
Phenotype
MIM: 102480 MIM: 606230
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACR PubMed Links

Gene Details

Gene
ACR
Gene Name
acrosin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001097.2 Intron NP_001088.2
Gene
LOC105373100
Gene Name
uncharacterized LOC105373100
There are no transcripts associated with this gene.

Gene
SHANK3
Gene Name
SH3 and multiple ankyrin repeat domains 3
There are no transcripts associated with this gene.

View Full Product Details