Product Details

SNP ID
rs80277373
Assay Type
Functionally tested
NCBI dbSNP Submissions
24
Location
Chr.1:12716158 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGCGCACAGCTTCCTCTCAGCCC[A/G]CTCTGAGCTGGAAGCAGCATGTGGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AADACL3 PubMed Links

Gene Details

Gene
AADACL3
Gene Name
arylacetamide deacetylase like 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001103170.2 44 UTR 5 NP_001096640.2

View Full Product Details