Product Details
- SNP ID
-
rs75727339
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:56338329 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TATAATCTTCTTTAATATAAAATAA[A/G]TCCACATGATGTATAATGGTAAAAT
- Phenotype
-
MIM: 614365
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
AASDH
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs35451510] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- AASDH
- Gene Name
- aminoadipate-semialdehyde dehydrogenase
- Gene
- KIAA1211
- Gene Name
- KIAA1211
There are no transcripts associated with this gene.
View Full Product Details