Product Details

SNP ID
rs77234282
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:107871160 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTATATCATGTGGAATGTGAGAGAA[C/T]GCCAACTTATGACCAAGATAAGTAA
Phenotype
MIM: 608648
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SEC63 PubMed Links

Gene Details

Gene
SEC63
Gene Name
SEC63 homolog, protein translocation regulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007214.4 2845 UTR 3 NP_009145.1
XM_011535399.2 2845 UTR 3 XP_011533701.1
XM_017010218.1 2845 UTR 3 XP_016865707.1

View Full Product Details