Product Details

SNP ID
rs76710364
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:124967362 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACAGAATGGTACTATGTTCATTT[C/T]AGAAATCTTCCACCAATATTTTTCT
Phenotype
MIM: 602532
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
RNF217 PubMed Links

Gene Details

Gene
RNF217
Gene Name
ring finger protein 217
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286398.2 1668 Intron NP_001273327.1
NM_152553.4 1668 Intron NP_689766.1
XM_011535494.2 1668 Intron XP_011533796.1
XM_011535495.2 1668 Intron XP_011533797.1
XM_011535496.2 1668 Intron XP_011533798.1
XM_011535499.2 1668 UTR 3 XP_011533801.1
XM_011535500.2 1668 Intron XP_011533802.1
XM_017010317.1 1668 UTR 3 XP_016865806.1
Gene
RNF217-AS1
Gene Name
RNF217 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details