Product Details
- SNP ID
-
rs79483955
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:138169836 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCTGTTCTTAGCCTTGAGTCTCTGA[C/T]ATATGTATATATTTCACATTTGTTA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ARFGEF3
PubMed Links
Gene Details
- Gene
- ARFGEF3
- Gene Name
- ARFGEF family member 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_020340.4 |
|
Intron |
|
|
NP_065073.3 |
View Full Product Details