Product Details

SNP ID
rs76496435
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:20197882 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACCAGACGTCTCTGTACCAATCTT[A/G]TGGTCAATTGCTGGGTACTTGGTTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR11G2 PubMed Links

Gene Details

Gene
OR11G2
Gene Name
olfactory receptor family 11 subfamily G member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005503.1 547 Missense Mutation ATG,GTG M183V NP_001005503.1

View Full Product Details