Product Details

SNP ID
rs78470960
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:56596962 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAATGCTCCACCTCACCTGGCAC[C/T]CCCACCCCCCAATGGAACATTTCTT
Phenotype
MIM: 605682 MIM: 602387
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BAZ2A PubMed Links

Gene Details

Gene
BAZ2A
Gene Name
bromodomain adjacent to zinc finger domain 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300905.1 7577 UTR 3 NP_001287834.1
NM_013449.3 7577 UTR 3 NP_038477.2
XM_011537797.2 7577 Intron XP_011536099.1
XM_011537798.2 7577 Intron XP_011536100.1
XM_011537799.2 7577 Intron XP_011536101.1
XM_011537800.2 7577 Intron XP_011536102.1
XM_011537801.2 7577 Intron XP_011536103.1
XM_011537803.2 7577 Intron XP_011536105.1
XM_011537804.2 7577 Intron XP_011536106.1
XM_011537805.2 7577 Intron XP_011536107.1
XM_011537806.2 7577 Intron XP_011536108.1
XM_011537807.2 7577 Intron XP_011536109.1
XM_011537809.2 7577 Intron XP_011536111.1
XM_017018740.1 7577 Intron XP_016874229.1
XM_017018741.1 7577 Intron XP_016874230.1
XM_017018742.1 7577 Intron XP_016874231.1
Gene
RBMS2
Gene Name
RNA binding motif single stranded interacting protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002898.3 7577 Intron NP_002889.1
XM_005269059.4 7577 Intron XP_005269116.1
XM_005269060.4 7577 Intron XP_005269117.1
XM_005269061.3 7577 Intron XP_005269118.1
XM_005269066.4 7577 Intron XP_005269123.1
XM_006719541.3 7577 Intron XP_006719604.1
XM_006719542.3 7577 Intron XP_006719605.1
XM_006719543.3 7577 Intron XP_006719606.1
XM_006719544.3 7577 Intron XP_006719607.1
XM_011538637.2 7577 Intron XP_011536939.1
XM_011538638.2 7577 Intron XP_011536940.1
XM_011538639.2 7577 Intron XP_011536941.1
XM_011538640.2 7577 Intron XP_011536942.1
XM_011538642.2 7577 Intron XP_011536944.1
XM_017019775.1 7577 Intron XP_016875264.1
XM_017019776.1 7577 Intron XP_016875265.1
XM_017019777.1 7577 Intron XP_016875266.1
XM_017019778.1 7577 Intron XP_016875267.1

View Full Product Details