Product Details

SNP ID
rs79133413
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:46033527 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAGATTCTTTGGCAGTTGTCTTT[T/G]TCATAACCCACAGGTGTAGAAACAC
Phenotype
MIM: 157145 MIM: 601984
Polymorphism
T/G, Transversion substitution
Allele Nomenclature
Literature Links
MSMB PubMed Links

Gene Details

Gene
MSMB
Gene Name
microseminoprotein beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002443.3 166 Missense Mutation GAA,GAC E80D NP_002434.1
NM_138634.2 166 Missense Mutation AAA,ACA K45T NP_619540.1
Gene
NCOA4
Gene Name
nuclear receptor coactivator 4
There are no transcripts associated with this gene.

View Full Product Details