Product Details

SNP ID
rs77922954
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:126017294 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCGGCTTCTCACTTAATATAGGC[A/G]GTGTGGGTGGATCTGGGCACTTGGT
Phenotype
MIM: 602714 MIM: 611640
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADAM12 PubMed Links

Gene Details

Gene
ADAM12
Gene Name
ADAM metallopeptidase domain 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001288973.1 3119 Silent Mutation ACC,ACT T902T NP_001275902.1
NM_001288974.1 3119 Intron NP_001275903.1
NM_001288975.1 3119 Intron NP_001275904.1
NM_003474.5 3119 Silent Mutation ACC,ACT T905T NP_003465.3
NM_021641.4 3119 Intron NP_067673.2
XM_017016705.1 3119 Silent Mutation ACC,ACT T749T XP_016872194.1
XM_017016706.1 3119 Silent Mutation ACC,ACT T516T XP_016872195.1
Gene
FANK1
Gene Name
fibronectin type III and ankyrin repeat domains 1
There are no transcripts associated with this gene.

View Full Product Details