Product Details

SNP ID
rs76400881
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:123023984 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCATGCCCATCAGTTGCTTGGAGC[A/T]CTATGGAGTGCACCACTGTCTGGTG
Phenotype
MIM: 123842
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
PPIC PubMed Links

Gene Details

Gene
PPIC
Gene Name
peptidylprolyl isomerase C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000943.4 635 Missense Mutation GAG,GTG E177V NP_000934.1
Gene
SNX24
Gene Name
sorting nexin 24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014035.2 635 Intron NP_054754.1
XM_005271972.4 635 Intron XP_005272029.1
XM_006714592.3 635 Intron XP_006714655.1
XM_011543349.2 635 Intron XP_011541651.1
XM_011543350.2 635 Intron XP_011541652.1
XM_011543352.1 635 Intron XP_011541654.1
XM_017009394.1 635 Intron XP_016864883.1
XM_017009395.1 635 Intron XP_016864884.1
XM_017009396.1 635 Intron XP_016864885.1

View Full Product Details