Product Details

SNP ID
rs75445510
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:47147478 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGCAGCCCCCGAAGGCCCTATCA[C/G]GACATGGAGTATGAAAGACGGTGAG
Phenotype
MIM: 300403 MIM: 300080
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
NDUFB11 PubMed Links

Gene Details

Gene
NDUFB11
Gene Name
NADH:ubiquinone oxidoreductase subunit B11
There are no transcripts associated with this gene.

Gene
RBM10
Gene Name
RNA binding motif protein 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204466.1 159 UTR 5 NP_001191395.1
NM_001204467.1 159 UTR 5 NP_001191396.1
NM_001204468.1 159 Missense Mutation CAC,CAG H64Q NP_001191397.1
NM_005676.4 159 UTR 5 NP_005667.2
NM_152856.2 159 UTR 5 NP_690595.1
XM_005272677.4 159 Missense Mutation CAC,CAG H64Q XP_005272734.1
XM_005272678.4 159 Missense Mutation CAC,CAG H64Q XP_005272735.1
XM_005272679.4 159 Missense Mutation CAC,CAG H64Q XP_005272736.1
XM_006724563.1 159 Intron XP_006724626.1
XM_011543989.2 159 Intron XP_011542291.1
XM_017029884.1 159 UTR 5 XP_016885373.1
XM_017029885.1 159 UTR 5 XP_016885374.1

View Full Product Details