Product Details

SNP ID
rs78340277
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:56700782 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCATCTCCAAGCCCCTGCTTTATG[C/T]CCAGGCCATGTCCATAAAGCTGTGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR9G1 PubMed Links

Gene Details

Gene
OR9G1
Gene Name
olfactory receptor family 9 subfamily G member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005213.1 395 Missense Mutation GCC,GTC A132V NP_001005213.1

View Full Product Details