Product Details

SNP ID
rs113289861
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:218664094 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGCGCTCCCAGCGAACACAGTCCC[A/G]AGTCCTGCGGGGTGGGGGCCCCTGC
Phenotype
MIM: 603647 MIM: 616014 MIM: 607652 MIM: 604083
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BCS1L PubMed Links

Gene Details

Gene
BCS1L
Gene Name
BCS1 homolog, ubiquinol-cytochrome c reductase complex chaperone
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001079866.1 1331 Intron NP_001073335.1
NM_001257342.1 1331 Intron NP_001244271.1
NM_001257343.1 1331 Intron NP_001244272.1
NM_001257344.1 1331 Intron NP_001244273.1
NM_001318836.1 1331 Intron NP_001305765.1
NM_001320717.1 1331 Intron NP_001307646.1
NM_004328.4 1331 Intron NP_004319.1
XM_005246748.2 1331 Intron XP_005246805.1
XM_005246749.3 1331 Intron XP_005246806.1
XM_006712678.1 1331 Intron XP_006712741.1
XM_017004631.1 1331 Intron XP_016860120.1
XM_017004632.1 1331 Intron XP_016860121.1
XM_017004633.1 1331 Intron XP_016860122.1
XM_017004634.1 1331 Intron XP_016860123.1
Gene
RNF25
Gene Name
ring finger protein 25
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022453.2 1331 Missense Mutation NP_071898.2
XM_017004695.1 1331 Missense Mutation XP_016860184.1
Gene
STK36
Gene Name
serine/threonine kinase 36
There are no transcripts associated with this gene.

Gene
ZNF142
Gene Name
zinc finger protein 142
There are no transcripts associated with this gene.

View Full Product Details