Product Details

SNP ID
rs115446532
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:241551159 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTCTGATCTGGAAACCAGGGCAAT[A/G]ACCTCATCCAGGGGCCTGGGCCAAG
Phenotype
MIM: 605404
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BOK PubMed Links

Gene Details

Gene
BOK
Gene Name
BOK, BCL2 family apoptosis regulator
There are no transcripts associated with this gene.

Gene
BOK-AS1
Gene Name
BOK antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details