Product Details

SNP ID
rs114266469
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:129252354 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCCTCACCAAGATTCTTTATCTCA[C/T]AAACCAGGTAACAGGGTGAAGCTTG
Phenotype
MIM: 615525
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
COPG1 PubMed Links

Gene Details

Gene
COPG1
Gene Name
coatomer protein complex subunit gamma 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016128.3 264 Missense Mutation ACA,ATA T55I NP_057212.1
XM_011512549.2 264 Missense Mutation ACA,ATA T55I XP_011510851.1

View Full Product Details