Product Details

SNP ID
rs115870489
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:25705236 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCAAGCAAGCTCTAAGACAGCCTC[C/T]CAGCGACATGAGACCCAAGGAGATA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SEL1L3 PubMed Links

Gene Details

Gene
SEL1L3
Gene Name
SEL1L family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297592.1 Intron NP_001284521.1
NM_001297594.1 Intron NP_001284523.1
NM_015187.4 Intron NP_056002.2
XM_011513819.2 Intron XP_011512121.2
XM_017007923.1 Intron XP_016863412.1

View Full Product Details