Product Details

SNP ID
rs117617524
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:83198864 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATATTCTTAAATGGTTATTTGGAA[A/G]ATGTGCTCATTTATAAAAGTTACAG
Phenotype
MIM: 172100
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PGM3 PubMed Links

Gene Details

Gene
PGM3
Gene Name
phosphoglucomutase 3
There are no transcripts associated with this gene.

Gene
RWDD2A
Gene Name
RWD domain containing 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322335.1 3523 UTR 3 NP_001309264.1
NM_001322336.1 3523 UTR 3 NP_001309265.1
NM_001322337.1 3523 UTR 3 NP_001309266.1
NM_001322339.1 3523 UTR 3 NP_001309268.1
NM_033411.4 3523 UTR 3 NP_219479.2
XM_017010222.1 3523 UTR 3 XP_016865711.1
XM_017010223.1 3523 UTR 3 XP_016865712.1

View Full Product Details