Product Details
- SNP ID
-
rs114778877
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:35795240 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTTGTCTCCCTCACAGGTCAGGCCA[C/T]GCTCACAGGGACACTTGTAGTAAAT
- Phenotype
-
MIM: 120105
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CLPS
PubMed Links
Gene Details
- Gene
- CLPS
- Gene Name
- colipase
- Gene
- CLPSL1
- Gene Name
- colipase like 1
There are no transcripts associated with this gene.
View Full Product Details