Product Details

SNP ID
rs117379990
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:41906859 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGACAGGCCAAATCCAGTAAGGCA[C/T]GGAGTAAAACAGCTGATGGGGGGCT
Phenotype
MIM: 612915
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MED20 PubMed Links

Gene Details

Gene
MED20
Gene Name
mediator complex subunit 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001305455.1 1098 UTR 3 NP_001292384.1
NM_001305456.1 1098 UTR 3 NP_001292385.1
NM_001305457.1 1098 UTR 3 NP_001292386.1
NM_004275.4 1098 UTR 3 NP_004266.2

View Full Product Details