Product Details

SNP ID
rs111764525
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:1541764 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGAGGCACCAGTGGGCTTGGCACT[C/T]CACCTTGGGCTTCCTTTTCCTGGAG
Phenotype
MIM: 600197
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC100128653 PubMed Links

Gene Details

Gene
LOC100128653
Gene Name
uncharacterized LOC100128653
There are no transcripts associated with this gene.

Gene
MAFK
Gene Name
MAF bZIP transcription factor K
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002360.3 2124 UTR 3 NP_002351.1
XM_005249851.2 2124 Intron XP_005249908.2
XM_006715773.2 2124 UTR 3 XP_006715836.1
Gene
TMEM184A
Gene Name
transmembrane protein 184A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001097620.1 2124 Intron NP_001091089.1
XM_006715653.1 2124 Intron XP_006715716.1
XM_006715655.2 2124 Intron XP_006715718.1
XM_011515164.1 2124 Intron XP_011513466.1
XM_011515165.2 2124 Intron XP_011513467.1
XM_011515166.2 2124 Intron XP_011513468.1
XM_017011815.1 2124 Intron XP_016867304.1

View Full Product Details