Product Details

SNP ID
rs115165773
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:87331288 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCCCCTAACTGCCTGTCTGACTG[G/T]TTTCTTCCTTTCTTCAATCTCAGTG
Phenotype
MIM: 616403
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TP53TG1 PubMed Links

Gene Details

Gene
TP53TG1
Gene Name
TP53 target 1 (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details