Product Details

SNP ID
rs113092320
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:26841143 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCAGCTAATAAAACACAATCCCT[A/G]CAACTTTTATATATAAGGAAAGTCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CAAP1 PubMed Links

Gene Details

Gene
CAAP1
Gene Name
caspase activity and apoptosis inhibitor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167575.1 2511 UTR 3 NP_001161047.1
NM_024828.3 2511 UTR 3 NP_079104.3

View Full Product Details