Product Details

SNP ID
rs114750615
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:13010933 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCGGATGGGGCTCCTGGCTCTGGC[A/G]GTCCCTCGCGGGAACCTGGGCCTCA
Phenotype
MIM: 614713
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RASSF10 PubMed Links

Gene Details

Gene
RASSF10
Gene Name
Ras association domain family member 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080521.2 1511 Missense Mutation AGT,GGT S453G NP_001073990.2

View Full Product Details