Product Details

SNP ID
rs114085531
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:13011750 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGAAGTCTAACCCTGTATTTCCTCC[G/T]AGTAAACACAATAGATGTTAAAAGG
Phenotype
MIM: 614713
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
RASSF10 PubMed Links

Gene Details

Gene
RASSF10
Gene Name
Ras association domain family member 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080521.2 2328 UTR 3 NP_001073990.2

View Full Product Details