Product Details

SNP ID
rs116462112
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:7989086 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTTTAAGCTCTACCTACCTTCCA[C/T]GTTCCCAGCAACAGCATTAAGTGCC
Phenotype
MIM: 603914
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EIF3F PubMed Links

Gene Details

Gene
EIF3F
Gene Name
eukaryotic translation initiation factor 3 subunit F
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003754.2 Intron NP_003745.1

View Full Product Details