Product Details

SNP ID
rs117799897
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:9470720 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGGGATTTTACTCTGAGAAAGTC[C/T]TCAGAAGGTTTTGAGCAGAAGCGGG
Phenotype
MIM: 603433
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF143 PubMed Links

Gene Details

Gene
ZNF143
Gene Name
zinc finger protein 143
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282656.1 Intron NP_001269585.1
NM_001282657.1 Intron NP_001269586.1
NM_003442.5 Intron NP_003433.3
XM_005253121.1 Intron XP_005253178.1
XM_005253122.1 Intron XP_005253179.1
XM_005253127.1 Intron XP_005253184.1
XM_011520349.2 Intron XP_011518651.1
XM_017018252.1 Intron XP_016873741.1
XM_017018253.1 Intron XP_016873742.1
XM_017018254.1 Intron XP_016873743.1
XM_017018255.1 Intron XP_016873744.1
XM_017018256.1 Intron XP_016873745.1

View Full Product Details