Product Details

SNP ID
rs114165962
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:31963542 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAAGCAGCTTCAAGTTGTTCAGAG[C/T]ATGCCTTTTGTAACTATAGTTAGAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KIAA1551 PubMed Links

Gene Details

Gene
KIAA1551
Gene Name
KIAA1551
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018169.3 Intron NP_060639.3
XM_005253405.2 Intron XP_005253462.1
XM_006719099.2 Intron XP_006719162.1
XM_011520722.1 Intron XP_011519024.1
XM_017019550.1 Intron XP_016875039.1

View Full Product Details