Product Details

SNP ID
rs116772643
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:10120406 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGGAATAATATATTTGAGTGGCT[A/G]AGAGGAAAAACAACAAACCTCTAAT
Phenotype
MIM: 606264
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CLEC7A PubMed Links

Gene Details

Gene
CLEC7A
Gene Name
C-type lectin domain family 7 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022570.4 Intron NP_072092.2
NM_197947.2 Intron NP_922938.1
NM_197948.2 Intron NP_922939.1
NM_197949.2 Intron NP_922940.1
NM_197950.2 Intron NP_922941.1
NM_197954.2 Intron NP_922945.1
XM_006719135.3 Intron XP_006719198.1
XM_017019822.1 Intron XP_016875311.1
XM_017019823.1 Intron XP_016875312.1

View Full Product Details