Product Details

SNP ID
rs116986462
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:18806534 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAAAAATAATCAATCAAACTTCAA[A/G]ACACACCAAAGCAACAGTAAAAGAG
Phenotype
MIM: 607669 MIM: 607032
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARL6IP1 PubMed Links

Gene Details

Gene
ARL6IP1
Gene Name
ADP ribosylation factor like GTPase 6 interacting protein 1
There are no transcripts associated with this gene.

Gene
SMG1
Gene Name
SMG1, nonsense mediated mRNA decay associated PI3K related kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015092.4 12341 UTR 3 NP_055907.3
XM_005255182.3 12341 UTR 3 XP_005255239.1
XM_005255183.3 12341 UTR 3 XP_005255240.1
XM_005255184.3 12341 UTR 3 XP_005255241.1
XM_011545769.2 12341 UTR 3 XP_011544071.1
XM_011545770.1 12341 UTR 3 XP_011544072.1
XM_017023066.1 12341 UTR 3 XP_016878555.1
XM_017023067.1 12341 UTR 3 XP_016878556.1

View Full Product Details