Product Details

SNP ID
rs118134901
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:27318595 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCAAGAAGTGCTGATGTCTCTTA[C/T]GTTTCAAGTCTAGCTCTGACTGACT
Phenotype
MIM: 147781
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IL4R PubMed Links

Gene Details

Gene
IL4R
Gene Name
interleukin 4 receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000418.3 Intron NP_000409.1
NM_001257406.1 Intron NP_001244335.1
NM_001257407.1 Intron NP_001244336.1
NM_001257997.1 Intron NP_001244926.1
XM_005255308.2 Intron XP_005255365.1
XM_006721043.1 Intron XP_006721106.1
XM_011545825.1 Intron XP_011544127.1
XM_011545826.2 Intron XP_011544128.1
XM_011545827.2 Intron XP_011544129.1
XM_011545828.2 Intron XP_011544130.1
XM_011545829.1 Intron XP_011544131.1
XM_011545830.2 Intron XP_011544132.1
XM_011545831.1 Intron XP_011544133.1
XM_011545832.1 Intron XP_011544134.1
XM_011545833.2 Intron XP_011544135.1
XM_011545834.2 Intron XP_011544136.1
XM_017023211.1 Intron XP_016878700.1

View Full Product Details