Product Details

SNP ID
rs117334934
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:66430845 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGTTCACTATATCCTCAGAGCCT[A/G]GACTAGTACCCAGCACTTATTGTGC
Phenotype
MIM: 612051
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BEAN1 PubMed Links

Gene Details

Gene
BEAN1
Gene Name
brain expressed, associated with NEDD4, 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136106.4 Intron NP_001129578.1
NM_001178020.2 Intron NP_001171491.1
NM_001197224.3 Intron NP_001184153.1
NM_001197225.3 Intron NP_001184154.1
XM_011522883.1 Intron XP_011521185.1
XM_011522884.1 Intron XP_011521186.1
XM_011522885.2 Intron XP_011521187.1
XM_011522887.2 Intron XP_011521189.1
XM_011522888.1 Intron XP_011521190.1
XM_011522889.1 Intron XP_011521191.1
XM_011522890.1 Intron XP_011521192.1
XM_011522891.1 Intron XP_011521193.1
XM_011522892.2 Intron XP_011521194.1
XM_011522894.2 Intron XP_011521196.1
XM_011522895.1 Intron XP_011521197.1

View Full Product Details