Product Details

SNP ID
rs112486552
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:8075702 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCACTCGGGTAATAATAGGTGATG[A/G]TCTCCGTCACATACCTGACCAGGGG
Phenotype
MIM: 603741
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ALOX12B PubMed Links

Gene Details

Gene
ALOX12B
Gene Name
arachidonate 12-lipoxygenase, 12R type
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001139.2 1808 Missense Mutation ACC,ATC T516I NP_001130.1

View Full Product Details