Product Details

SNP ID
rs116832595
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:8753873 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGTGTCCTGCCATCCTTATACAA[A/G]ACAGATGACGTCACCTCATCCTAGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC42 PubMed Links

Gene Details

Gene
CCDC42
Gene Name
coiled-coil domain containing 42
There are no transcripts associated with this gene.

Gene
SPDYE4
Gene Name
speedy/RINGO cell cycle regulator family member E4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128076.1 Intron NP_001121548.1
XM_005256632.4 Intron XP_005256689.1
XM_005256633.4 Intron XP_005256690.1

View Full Product Details