Product Details

SNP ID
rs113664821
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:39868356 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTGCGACGCCAGCCCCTGAGCGA[C/T]GATGCGAACGTGCGTCCTACTCTCC
Phenotype
MIM: 606221 MIM: 608499
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IKZF3 PubMed Links

Gene Details

Gene
IKZF3
Gene Name
IKAROS family zinc finger 3
There are no transcripts associated with this gene.

Gene
ZPBP2
Gene Name
zona pellucida binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198844.2 155 Missense Mutation ACG,ATG T1M NP_942141.2
NM_199321.2 155 Missense Mutation ACG,ATG T1M NP_955353.1
XM_011524298.1 155 Missense Mutation ACG,ATG T1M XP_011522600.1

View Full Product Details