Product Details
- SNP ID
-
rs115451635
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:81035271 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCAGGACCATGTCTCTGTCTCGCT[C/T]AGAGGAGATGCACCGGCTCACGGAA
- Phenotype
-
MIM: 605475
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
BAIAP2
PubMed Links
Gene Details
- Gene
- BAIAP2
- Gene Name
- BAI1 associated protein 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001144888.1 |
125 |
Missense Mutation |
TCA,TTA |
S6L |
NP_001138360.1 |
NM_006340.2 |
125 |
Missense Mutation |
TCA,TTA |
S6L |
NP_006331.1 |
NM_017450.2 |
125 |
Missense Mutation |
TCA,TTA |
S6L |
NP_059344.1 |
NM_017451.2 |
125 |
Missense Mutation |
TCA,TTA |
S6L |
NP_059345.1 |
XM_005256943.1 |
125 |
Missense Mutation |
TCA,TTA |
S6L |
XP_005257000.1 |
XM_005256944.1 |
125 |
Missense Mutation |
TCA,TTA |
S6L |
XP_005257001.1 |
XM_005256945.1 |
125 |
Missense Mutation |
TCA,TTA |
S6L |
XP_005257002.1 |
XM_005256948.3 |
125 |
Missense Mutation |
TCA,TTA |
S6L |
XP_005257005.1 |
XM_006721635.1 |
125 |
Missense Mutation |
TCA,TTA |
S6L |
XP_006721698.1 |
XM_006721636.1 |
125 |
Missense Mutation |
TCA,TTA |
S6L |
XP_006721699.1 |
XM_006721637.2 |
125 |
Missense Mutation |
TCA,TTA |
S6L |
XP_006721700.1 |
XM_011524193.1 |
125 |
Missense Mutation |
TCA,TTA |
S6L |
XP_011522495.1 |
XM_011524194.1 |
125 |
Missense Mutation |
TCA,TTA |
S6L |
XP_011522496.1 |
XM_011524195.1 |
125 |
Intron |
|
|
XP_011522497.1 |
XM_011524196.1 |
125 |
Intron |
|
|
XP_011522498.1 |
XM_011524197.1 |
125 |
Intron |
|
|
XP_011522499.1 |
XM_017024017.1 |
125 |
Intron |
|
|
XP_016879506.1 |
XM_017024018.1 |
125 |
Intron |
|
|
XP_016879507.1 |
XM_017024019.1 |
125 |
Intron |
|
|
XP_016879508.1 |
- Gene
- BAIAP2-AS1
- Gene Name
- BAIAP2 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.
View Full Product Details