Product Details

SNP ID
rs115565555
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:29561636 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCCAGTAGTAGGATCAGAAAAGG[G/T]AGCAACCAACCAGAAAACACCTAAG
Phenotype
MIM: 610266
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ABHD15 PubMed Links

Gene Details

Gene
ABHD15
Gene Name
abhydrolase domain containing 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198147.2 2390 UTR 3 NP_937790.2
Gene
TAOK1
Gene Name
TAO kinase 1
There are no transcripts associated with this gene.

Gene
TP53I13
Gene Name
tumor protein p53 inducible protein 13
There are no transcripts associated with this gene.

View Full Product Details