Product Details
- SNP ID
-
rs114402161
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:12993710 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGCCTAGGTGGGAATCCTCTGGCCT[C/T]ATTTGTGCTATGTGGACTGGAAACG
- Phenotype
-
MIM: 616426
MIM: 609263
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CEP192
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs201371427] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CEP192
- Gene Name
- centrosomal protein 192
- Gene
- SEH1L
- Gene Name
- SEH1 like nucleoporin
There are no transcripts associated with this gene.
View Full Product Details