Product Details

SNP ID
rs117405702
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:46478520 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACCTGTGGTCAATTCTCTATGAAG[C/T]AGCCATGTGATCCTATTGAGAGGTA
Phenotype
MIM: 613072
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOXHD1 PubMed Links

Gene Details

Gene
LOXHD1
Gene Name
lipoxygenase homology domains 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145472.2 Intron NP_001138944.1
NM_001145473.2 Intron NP_001138945.1
NM_001173129.1 Intron NP_001166600.1
NM_001308013.1 Intron NP_001294942.1
NM_144612.6 Intron NP_653213.6
XM_006722388.3 Intron XP_006722451.1
XM_006722389.3 Intron XP_006722452.1
XM_006722390.3 Intron XP_006722453.1
XM_006722391.3 Intron XP_006722454.1
XM_011525804.2 Intron XP_011524106.1
XM_011525807.2 Intron XP_011524109.1
XM_011525810.2 Intron XP_011524112.1
XM_011525811.2 Intron XP_011524113.1
XM_017025540.1 Intron XP_016881029.1
XM_017025541.1 Intron XP_016881030.1
XM_017025542.1 Intron XP_016881031.1
XM_017025543.1 Intron XP_016881032.1
XM_017025544.1 Intron XP_016881033.1
XM_017025545.1 Intron XP_016881034.1
XM_017025546.1 Intron XP_016881035.1
XM_017025547.1 Intron XP_016881036.1
XM_017025548.1 Intron XP_016881037.1

View Full Product Details