Product Details
- SNP ID
-
rs112126725
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:39296443 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGCCATGGCTGCAGCTTGGACCGTG[A/G]TGCTGGTGACTTTGGTGCTAGGCTT
- Phenotype
-
MIM: 607403
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
IFNL1
PubMed Links
Gene Details
- Gene
- IFNL1
- Gene Name
- interferon lambda 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_172140.1 |
119 |
Missense Mutation |
ATG,GTG |
M8V |
NP_742152.1 |
View Full Product Details