Product Details
- SNP ID
-
rs117228099
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:39784670 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTCAAAGCTACAACTTGATCTATCC[A/G]TAGTAAAGGTCTGTTCCCAAGTGTG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
LEUTX
PubMed Links
Gene Details
- Gene
- LEUTX
- Gene Name
- leucine twenty homeobox
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001143832.1 |
226 |
Missense Mutation |
ATA,GTA |
I21V |
NP_001137304.1 |
View Full Product Details