Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001005851.2 | 7580 | UTR 3 | NP_001005851.1 | ||
XM_005258590.4 | 7580 | Intron | XP_005258647.1 | ||
XM_005258591.4 | 7580 | Intron | XP_005258648.1 | ||
XM_005258592.3 | 7580 | Intron | XP_005258649.1 | ||
XM_005258593.2 | 7580 | Intron | XP_005258650.1 | ||
XM_005258595.3 | 7580 | Intron | XP_005258652.1 | ||
XM_006723072.3 | 7580 | Intron | XP_006723135.1 | ||
XM_011526593.2 | 7580 | Intron | XP_011524895.1 | ||
XM_017026425.1 | 7580 | Intron | XP_016881914.1 | ||
XM_017026426.1 | 7580 | Intron | XP_016881915.1 | ||
XM_017026427.1 | 7580 | Intron | XP_016881916.1 |