Product Details

SNP ID
rs116472187
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:40741667 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGCCAGGAGACAGCCTGGCTGAGA[C/T]ACTGACTACCTAGCTTGAATCCTGA
Phenotype
MIM: 606476 MIM: 182285
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C19orf54 PubMed Links

Gene Details

Gene
C19orf54
Gene Name
chromosome 19 open reading frame 54
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198476.3 1482 UTR 3 NP_940878.3
XM_005258776.4 1482 UTR 3 XP_005258833.1
XM_005258778.4 1482 UTR 3 XP_005258835.1
XM_006723152.2 1482 Intron XP_006723215.1
XM_006723153.2 1482 Intron XP_006723216.1
XM_011526774.2 1482 UTR 3 XP_011525076.1
XM_011526775.1 1482 Intron XP_011525077.1
XM_011526776.2 1482 UTR 3 XP_011525078.1
XM_011526777.2 1482 UTR 3 XP_011525079.1
XM_011526778.2 1482 UTR 3 XP_011525080.1
XM_011526779.2 1482 Intron XP_011525081.1
XM_011526782.2 1482 Intron XP_011525084.1
XM_011526783.2 1482 UTR 3 XP_011525085.1
XM_011526784.2 1482 UTR 3 XP_011525086.1
XM_011526785.2 1482 UTR 3 XP_011525087.1
XM_011526786.2 1482 Intron XP_011525088.1
XM_017026620.1 1482 Intron XP_016882109.1
XM_017026621.1 1482 Intron XP_016882110.1
XM_017026622.1 1482 Intron XP_016882111.1
XM_017026623.1 1482 UTR 3 XP_016882112.1
XM_017026624.1 1482 Intron XP_016882113.1
XM_017026625.1 1482 Intron XP_016882114.1
XM_017026626.1 1482 UTR 3 XP_016882115.1
XM_017026627.1 1482 UTR 3 XP_016882116.1
Gene
ITPKC
Gene Name
inositol-trisphosphate 3-kinase C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025194.2 1482 Intron NP_079470.1
XM_017027324.1 1482 Intron XP_016882813.1
Gene
SNRPA
Gene Name
small nuclear ribonucleoprotein polypeptide A
There are no transcripts associated with this gene.

View Full Product Details