Product Details

SNP ID
rs116144364
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:40742378 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCCATCACACAGTGGGAGAAGTGT[A/G]GGCACACTTCAAATGGTTACCGAGC
Phenotype
MIM: 606476 MIM: 182285
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C19orf54 PubMed Links

Gene Details

Gene
C19orf54
Gene Name
chromosome 19 open reading frame 54
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198476.3 1687 UTR 3 NP_940878.3
XM_005258776.4 1687 UTR 3 XP_005258833.1
XM_005258778.4 1687 UTR 3 XP_005258835.1
XM_006723152.2 1687 Intron XP_006723215.1
XM_006723153.2 1687 Intron XP_006723216.1
XM_011526774.2 1687 Intron XP_011525076.1
XM_011526775.1 1687 Intron XP_011525077.1
XM_011526776.2 1687 Intron XP_011525078.1
XM_011526777.2 1687 Intron XP_011525079.1
XM_011526778.2 1687 Intron XP_011525080.1
XM_011526779.2 1687 Intron XP_011525081.1
XM_011526782.2 1687 Intron XP_011525084.1
XM_011526783.2 1687 Intron XP_011525085.1
XM_011526784.2 1687 Intron XP_011525086.1
XM_011526785.2 1687 Intron XP_011525087.1
XM_011526786.2 1687 Intron XP_011525088.1
XM_017026620.1 1687 Intron XP_016882109.1
XM_017026621.1 1687 Intron XP_016882110.1
XM_017026622.1 1687 Intron XP_016882111.1
XM_017026623.1 1687 Intron XP_016882112.1
XM_017026624.1 1687 Intron XP_016882113.1
XM_017026625.1 1687 Intron XP_016882114.1
XM_017026626.1 1687 UTR 3 XP_016882115.1
XM_017026627.1 1687 UTR 3 XP_016882116.1
Gene
ITPKC
Gene Name
inositol-trisphosphate 3-kinase C
There are no transcripts associated with this gene.

Gene
SNRPA
Gene Name
small nuclear ribonucleoprotein polypeptide A
There are no transcripts associated with this gene.

View Full Product Details